Learn More
Medchemexpress LLC PDGF-BB Rat 100ug
SDP
Catalog No. 5000181761
Encompass_Preferred
PDGF-BB Protein Rat is a member of PDGF family which promotes cell proliferation survival and migration through binding to the tyrosine kinase PDGF receptor
Catalog No. 50-001-81761 Supplier Medchemexpress LLC Supplier No. HYP7278100UG
PDGF-BB Protein Rat is a member of PDGF family which promotes cell proliferation survival and migration through binding to the tyrosine kinase PDGF receptor