Learn More
Medchemexpress LLC HY-P7278 50ug Medchemexpress, PDGF-BB, Rat CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP727850UG
Encompass_Preferred
Medchemexpress, HY-P7278 50ug PDGF-BB, Rat CAS: Recombinant Rat Platelet-Derived Growth Factor-BB is a member of PDGF family, which promotes cell proliferation, survival and migration, through binding to the tyrosine kinase PDGF receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4373