Learn More
Medchemexpress LLC PDGF-BB Rat His 10ug
SDP
Catalog No. 5000181665
Encompass_Preferred
PDGF-BB Protein Rat is a member of PDGF family which promotes cell proliferation survival and migration through binding to the tyrosine kinase PDGF receptor
Catalog No. 50-001-81665 Supplier Medchemexpress LLC Supplier No. HYP7278A10UG
PDGF-BB Protein Rat is a member of PDGF family which promotes cell proliferation survival and migration through binding to the tyrosine kinase PDGF receptor