Learn More
Medchemexpress LLC PDGF-BB Rat His 50ug
SDP
Catalog No. 5000181693
Encompass_Preferred
PDGF-BB Protein Rat is a member of PDGF family which promotes cell proliferation survival and migration through binding to the tyrosine kinase PDGF receptor
Catalog No. 50-001-81693 Supplier Medchemexpress LLC Supplier No. HYP7278A50UG
PDGF-BB Protein Rat is a member of PDGF family which promotes cell proliferation survival and migration through binding to the tyrosine kinase PDGF receptor