Learn More
Medchemexpress LLC PDGF-CC Mouse HEK2 50ug
SDP
Catalog No. 5000179193
Encompass_Preferred
The PDGF-CC protein is a key growth factor that plays crucial roles in a variety of cellular processes including embryonic development cell proliferation migration survival and chemotaxis It has potent mitogenic and chemoattractant properties and is essential during embryonic skeletal formation craniofacial development palate development skin morphogenesis and wound healing stages PDGF-CC Protein Mouse (HEK293 Fc) is the recombinant mouse-derived PDGF-CC protein expressed by HEK293 with N-hFc labeled tag
Catalog No. 50-001-79193 Supplier Medchemexpress LLC Supplier No. HYP7335450UG
The PDGF-CC protein is a key growth factor that plays crucial roles in a variety of cellular processes including embryonic development cell proliferation migration survival and chemotaxis It has potent mitogenic and chemoattractant properties and is essential during embryonic skeletal formation craniofacial development palate development skin morphogenesis and wound healing stages PDGF-CC Protein Mouse (HEK293 Fc) is the recombinant mouse-derived PDGF-CC protein expressed by HEK293 with N-hFc labeled tag