Learn More
Medchemexpress LLC HY-76210 500mg Medchemexpress, 5-Hydroxypyrazine-2-Carboxylic Acid CAS:34604-60-9 Purity:>98%
SDP
Catalog No. 501970080
Encompass_Preferred
Medchemexpress, HY-P7280 10ug PDGF-DD (CHO-expressed), Human CAS: Recombinant Human PDGF-DD (CHO-expressed) is a member of the PDGF family, specifically binds to and activates its cognate receptor PDGFR-β, and participates in regulating cancer proliferation, transformation, and invasion through activating multiple downstream oncogenic pathways, resulting in tumor development and progression. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0080 Supplier Medchemexpress LLC Supplier No. HY76210500MG
Medchemexpress, HY-P7280 10ug PDGF-DD (CHO-expressed), Human CAS: Recombinant Human PDGF-DD (CHO-expressed) is a member of the PDGF family, specifically binds to and activates its cognate receptor PDGFR-β, and participates in regulating cancer proliferation, transformation, and invasion through activating multiple downstream oncogenic pathways, resulting in tumor development and progression. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.