Learn More
Medchemexpress LLC PDGF-DD Human CHO 50ug
SDP
Catalog No. 5000182742
Encompass_Preferred
PDGF-DD Protein Human (CHO) is a member of the PDGF family specifically binds to and activates its cognate receptor PDGFR- and participates in regulating cancer proliferation transformation and invasion through activating multiple downstream oncogenic pathways resulting in tumor development and progression
Catalog No. 50-001-82742 Supplier Medchemexpress LLC Supplier No. HYP728050UG
PDGF-DD Protein Human (CHO) is a member of the PDGF family specifically binds to and activates its cognate receptor PDGFR- and participates in regulating cancer proliferation transformation and invasion through activating multiple downstream oncogenic pathways resulting in tumor development and progression