Learn More
Medchemexpress LLC Recombinant human platelet-derived growth factor D (PDGF-DD) | >95.0% | 50UG
SDP
Catalog No. 5000174062
Encompass_Preferred
Recombinant human platelet-derived growth factor D (PDGF-DD) is a glycosylated growth factor produced in HEK293 cells and supplied as an N-terminal His-tagged, lyophilized protein. It promotes cell proliferation, migration, and angiogenic responses through platelet-derived growth factor receptor activation and is characterized for research use with specified purity and endotoxin limits.
  • Expressed in HEK293 cells.
  • N-terminal His tag.
  • Lyophilized powder formulated in PBS with 8% trehalose.
  • Purity >95% by reducing SDS-PAGE.
  • Endotoxin <1 EU/μg (LAL method).
  • Do not reconstitute below 100 μg/mL; aliquot and freeze for long-term storage.
  • Available in microgram-scale sizes such as 50 μg.
Catalog No. 50-001-74062 Supplier Medchemexpress LLC Supplier No. HYP70283750UG
Recombinant human platelet-derived growth factor D (PDGF-DD) is a glycosylated growth factor produced in HEK293 cells and supplied as an N-terminal His-tagged, lyophilized protein. It promotes cell proliferation, migration, and angiogenic responses through platelet-derived growth factor receptor activation and is characterized for research use with specified purity and endotoxin limits.
  • Expressed in HEK293 cells.
  • N-terminal His tag.
  • Lyophilized powder formulated in PBS with 8% trehalose.
  • Purity >95% by reducing SDS-PAGE.
  • Endotoxin <1 EU/μg (LAL method).
  • Do not reconstitute below 100 μg/mL; aliquot and freeze for long-term storage.
  • Available in microgram-scale sizes such as 50 μg.