Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PDGF-DD Mouse His- 50ug
SDP

Catalog No. 5000196318
Encompass_Preferred

Recombinant mouse PDGF-DD is supplied lyophilized (50 μg) as an N-terminal SUMO and 6xHis tagged protein expressed in E. coli. The protein is purified to greater than 90% by reducing SDS-PAGE, formulated in 10 mM Tris-HCl, 1 mM EDTA, 6% trehalose (pH 8.0), and has an endotoxin level below 1 EU/μg. Store at -20°C; after reconstitution, the material is stable at 4°C for 1 week or at -20°C for longer-term storage-aliquot to avoid repeated freeze-thaw cycles.

  • Recombinant mouse PDGF-DD with N-terminal SUMO and 6xHis tag.
  • Expression in E. coli for high-yield production.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Low endotoxin content (<1 EU/μg) suitable for in vitro assays.
  • Lyophilized formulation with trehalose for improved stability.
  • Recommended storage at -20°C; aliquot to avoid freeze-thaw.

Catalog No. 50-001-96318 Supplier Medchemexpress LLC Supplier No. HYP7161550UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse PDGF-DD is supplied lyophilized (50 μg) as an N-terminal SUMO and 6xHis tagged protein expressed in E. coli. The protein is purified to greater than 90% by reducing SDS-PAGE, formulated in 10 mM Tris-HCl, 1 mM EDTA, 6% trehalose (pH 8.0), and has an endotoxin level below 1 EU/μg. Store at -20°C; after reconstitution, the material is stable at 4°C for 1 week or at -20°C for longer-term storage-aliquot to avoid repeated freeze-thaw cycles.

  • Recombinant mouse PDGF-DD with N-terminal SUMO and 6xHis tag.
  • Expression in E. coli for high-yield production.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Low endotoxin content (<1 EU/μg) suitable for in vitro assays.
  • Lyophilized formulation with trehalose for improved stability.
  • Recommended storage at -20°C; aliquot to avoid freeze-thaw.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.