Learn More
Genscript Corporation PDGF R alpha/PDGFRABiotin H
Supplier:  Genscript Corporation Z04583500
Encompass
Platelet-derived growth factor is commonly known as a mitogen Many research data suggest a role for PDGF-beta R in the mitogenic response of mesangial cells There are four members of PDGF family known as PDGF-A chain PDGF-B chain PDGF-C chain and PDGF-D chain which in active forms are dimers As far as two receptors PDGF-alpha R and PDGF-beta R are known to bind PDGF
Catalog No. 50-000-57633