Learn More
Genscript Corporation PDGF R beta/CD140bBiotin Hi
Catalog No. 5000056725
Encompass
PDGF is a major serum mitogen that can exist as a homo or hetero-dimeric protein consisting of disulfide-linked PDGF-A and PDGF-B chains The PDGF-AA PDGF-BB and PDGF-AB isoforms have been shown to bind to two distinct cell surface PDGF receptors with different affinities Where as PDGF R alpha binds all three PDGF isoforms with high affinity PDGF R beta binds PDGF-BB only with high-affinity PDGF R plays an essential role in blood vessel development by promoting proliferation migration and recruitment of pericytes and smooth muscle cells to endothelial cells Plays a role in the migration of vascular smooth muscle cells and the formation of neointima at vascular injury sites
Catalog No. 50-000-56725 Supplier Genscript Corporation Supplier No. Z04584100
PDGF is a major serum mitogen that can exist as a homo or hetero-dimeric protein consisting of disulfide-linked PDGF-A and PDGF-B chains The PDGF-AA PDGF-BB and PDGF-AB isoforms have been shown to bind to two distinct cell surface PDGF receptors with different affinities Where as PDGF R alpha binds all three PDGF isoforms with high affinity PDGF R beta binds PDGF-BB only with high-affinity PDGF R plays an essential role in blood vessel development by promoting proliferation migration and recruitment of pericytes and smooth muscle cells to endothelial cells Plays a role in the migration of vascular smooth muscle cells and the formation of neointima at vascular injury sites