Learn More
Medchemexpress LLC PDGF R beta Human 100ug
SDP
Catalog No. 5000182128
Encompass_Preferred
TRF1 protein is a telomere-binding protein that plays a crucial role in telomere maintenance and chromosome stability It is involved in regulating telomere length and preventing DNA damage PDGF R beta Protein Human (HEK293 His) is the recombinant human-derived PDGF R beta protein expressed by HEK293 with C-6 His labeled tag
Catalog No. 50-001-82128 Supplier Medchemexpress LLC Supplier No. HYP72494100UG
TRF1 protein is a telomere-binding protein that plays a crucial role in telomere maintenance and chromosome stability It is involved in regulating telomere length and preventing DNA damage PDGF R beta Protein Human (HEK293 His) is the recombinant human-derived PDGF R beta protein expressed by HEK293 with C-6 His labeled tag