Learn More
Medchemexpress LLC PDGF R beta Human 50ug | 50ug
SDP
Catalog No. 5000185679
Encompass_Preferred
PDGF R protein is integral in embryonic development and regulates cell proliferation survival differentiation chemotaxis migration and vascular development It crucially recruits pericytes and smooth muscle cells to the endothelium promoting vascular smooth muscle cell migration and neointimal formation at the site of injury PDGF R beta Protein Human (HEK293 C-His-Avi) is the recombinant human-derived PDGF R beta protein expressed by HEK293 with C-His C-Avi labeled tag
Catalog No. 50-001-85679 Supplier Medchemexpress LLC Supplier No. HYP7712350UG
PDGF R protein is integral in embryonic development and regulates cell proliferation survival differentiation chemotaxis migration and vascular development It crucially recruits pericytes and smooth muscle cells to the endothelium promoting vascular smooth muscle cell migration and neointimal formation at the site of injury PDGF R beta Protein Human (HEK293 C-His-Avi) is the recombinant human-derived PDGF R beta protein expressed by HEK293 with C-His C-Avi labeled tag