Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PDGF R beta protein, rat (HEK293, Fc) | >95.0% | 10 UG
SDP

Catalog No. 5000293365
Encompass_Preferred

Recombinant PDGF R beta protein, rat, is expressed in HEK293 cells with a C-terminal Fc tag and is supplied for use in receptor-ligand binding and cell-signaling studies. Purity is >95% as determined by reducing SDS-PAGE. The product is offered in microgram-scale sizes and the manufacturer recommends not reconstituting to a concentration less than 100 μg/mL in PBS.

  • Recombinant protein expressed in HEK293 cells.
  • C-terminal Fc tag for detection and purification.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Recommended reconstitution concentration ≥100 μg/mL in PBS.
  • Available in microgram-scale sizes suitable for small-scale assays.

Catalog No. 50-002-93365 Supplier Medchemexpress LLC Supplier No. HYP7464410UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant PDGF R beta protein, rat, is expressed in HEK293 cells with a C-terminal Fc tag and is supplied for use in receptor-ligand binding and cell-signaling studies. Purity is >95% as determined by reducing SDS-PAGE. The product is offered in microgram-scale sizes and the manufacturer recommends not reconstituting to a concentration less than 100 μg/mL in PBS.

  • Recombinant protein expressed in HEK293 cells.
  • C-terminal Fc tag for detection and purification.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Recommended reconstitution concentration ≥100 μg/mL in PBS.
  • Available in microgram-scale sizes suitable for small-scale assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.