Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PDGF R beta protein, rat (HEK293, Fc) | >95.0% | 100 UG
SDP

Catalog No. 5000293261
Encompass_Preferred

Recombinant rat platelet-derived growth factor receptor beta (PDGFR-β) extracellular construct (L32-K530) expressed in HEK293 cells and supplied as a C-terminal human Fc fusion. Supplied as a lyophilized powder for research use, with formulation and storage recommendations provided in the product documentation.

  • Contains amino acid construct L32-K530, corresponding to the extracellular region of PDGFR-β.
  • Expressed in HEK293 cells with a C-terminal human Fc fusion tag.
  • Lyophilized powder formulated from a 0.22 μm filtered PBS solution, pH 7.4.
  • Purity >95.0% as determined by reducing SDS-PAGE.
  • Available in multiple sizes, including 100 μg.
  • Intended for research use only; not for human or clinical use.
  • Reconstitute at ≥100 μg/mL in PBS; add carrier protein for long-term storage.

Catalog No. 50-002-93261 Supplier Medchemexpress LLC Supplier No. HYP74644100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant rat platelet-derived growth factor receptor beta (PDGFR-β) extracellular construct (L32-K530) expressed in HEK293 cells and supplied as a C-terminal human Fc fusion. Supplied as a lyophilized powder for research use, with formulation and storage recommendations provided in the product documentation.

  • Contains amino acid construct L32-K530, corresponding to the extracellular region of PDGFR-β.
  • Expressed in HEK293 cells with a C-terminal human Fc fusion tag.
  • Lyophilized powder formulated from a 0.22 μm filtered PBS solution, pH 7.4.
  • Purity >95.0% as determined by reducing SDS-PAGE.
  • Available in multiple sizes, including 100 μg.
  • Intended for research use only; not for human or clinical use.
  • Reconstitute at ≥100 μg/mL in PBS; add carrier protein for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.