Learn More
Medchemexpress LLC PDGF R beta Rat HE 10ug
SDP
Catalog No. 5000179943
Encompass_Preferred
PDGF R beta Protein a receptor tyrosine kinase binds ligands PDGFA PDGFB and PDGFD forming homodimers or heterodimers Ligand binding activates diverse signaling cascades in PDGF R beta with responses contingent on the specific ligand The modulation of responses involves heterodimer formation with another receptor PDGFRB PDGF R beta Protein Rat (HEK293 His) is the recombinant rat-derived PDGF R beta protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-79943 Supplier Medchemexpress LLC Supplier No. HYP7336010UG
PDGF R beta Protein a receptor tyrosine kinase binds ligands PDGFA PDGFB and PDGFD forming homodimers or heterodimers Ligand binding activates diverse signaling cascades in PDGF R beta with responses contingent on the specific ligand The modulation of responses involves heterodimer formation with another receptor PDGFRB PDGF R beta Protein Rat (HEK293 His) is the recombinant rat-derived PDGF R beta protein expressed by HEK293 with C-His labeled tag