Learn More
Medchemexpress LLC PDGF R beta Rhesus 2ug | 2ug
SDP
Catalog No. 5000185874
Encompass_Preferred
PDGF R beta a tyrosine-protein kinase receptor crucially regulates embryonic development overseeing cell processes like proliferation survival differentiation chemotaxis and migration Especially vital in blood vessel development it facilitates pericyte and smooth muscle cell proliferation migration and recruitment to endothelial cells playing a significant role in blood vessel formation and maintenance PDGF R beta Protein Rhesus Macaque (HEK293 His) is the recombinant Rhesus Macaque-derived PDGF R beta protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-85874 Supplier Medchemexpress LLC Supplier No. HYP771292UG
PDGF R beta a tyrosine-protein kinase receptor crucially regulates embryonic development overseeing cell processes like proliferation survival differentiation chemotaxis and migration Especially vital in blood vessel development it facilitates pericyte and smooth muscle cell proliferation migration and recruitment to endothelial cells playing a significant role in blood vessel formation and maintenance PDGF R beta Protein Rhesus Macaque (HEK293 His) is the recombinant Rhesus Macaque-derived PDGF R beta protein expressed by HEK293 with C-His labeled tag