Learn More
Medchemexpress LLC PDGF R beta Rhesus 500ug | 500ug
SDP
Catalog No. 5000185591
Encompass_Preferred
PDGF R beta a tyrosine-protein kinase receptor crucially regulates embryonic development overseeing cell processes like proliferation survival differentiation chemotaxis and migration Especially vital in blood vessel development it facilitates pericyte and smooth muscle cell proliferation migration and recruitment to endothelial cells playing a significant role in blood vessel formation and maintenance PDGF R beta Protein Rhesus Macaque (HEK293 His) is the recombinant Rhesus Macaque-derived PDGF R beta protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-85591 Supplier Medchemexpress LLC Supplier No. HYP77129500UG
PDGF R beta a tyrosine-protein kinase receptor crucially regulates embryonic development overseeing cell processes like proliferation survival differentiation chemotaxis and migration Especially vital in blood vessel development it facilitates pericyte and smooth muscle cell proliferation migration and recruitment to endothelial cells playing a significant role in blood vessel formation and maintenance PDGF R beta Protein Rhesus Macaque (HEK293 His) is the recombinant Rhesus Macaque-derived PDGF R beta protein expressed by HEK293 with C-His labeled tag