Learn More
Aviva Systems Biology Corp PDGFD Antibody - N-terminal region (ARP58814_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58814_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PDGFD. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PDGFD. Alternate names: IEGF, MGC26867, MSTP036, SCDGF-B, SCDGFBApplicationWB. Species reactivity: Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62433