Learn More
Eurofins Discoverx PDGFRα, active, 50 µg. (14-467)
Supplier:  Eurofins Discoverx 14467
Encompass
N-terminal 6xHis-tagged recombinant, human PDGFRα, amino acids 550-end. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8837