Learn More
Sino Biological Recombinant Human PCTK2 (CDK17) & Cyclin Y Protein (GST Tag), Active
Catalog No. 502573899
Encompass_Preferred
Recombinant full length human PCTK2 (CDK17) and Cyclin Y (2-end) were co-expressed by baculovirus in Sf9 insect cells using N-terminal GST tags.
Catalog No. 50-257-3899 Supplier Sino Biological Supplier No. P1010G50
Recombinant full length human PCTK2 (CDK17) and Cyclin Y (2-end) were co-expressed by baculovirus in Sf9 insect cells using N-terminal GST tags.