Learn More
Medchemexpress LLC HY-111507 10mg Medchemexpress, PDGFRα kinase inhibitor 1 CAS:2209053-93-8 Purity:>98%
SDP
Catalog No. 501968895
Encompass_Preferred
Medchemexpress, HY-111507 10mg PDGFRα kinase inhibitor 1 CAS:2209053-93-8 PDGFRα kinase inhibitor 1 is a highly selective type II PDGFRα kinase inhibitor with IC50s of 132 nM and 6115 nM for PDGFRα and PDGFRβ, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8895 Supplier Medchemexpress LLC Supplier No. HY11150710MG
Medchemexpress, HY-111507 10mg PDGFRα kinase inhibitor 1 CAS:2209053-93-8 PDGFRα kinase inhibitor 1 is a highly selective type II PDGFRα kinase inhibitor with IC50s of 132 nM and 6115 nM for PDGFRα and PDGFRβ, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.