Learn More
Medchemexpress LLC PDGFRα kinase inhibitor 1 | 2209053-93-8 | 98.5% | 586.69 | 50 MG
SDP
Catalog No. 5000344248
Encompass_Preferred
PDGFRα kinase inhibitor 1 is a highly selective type II PDGFRα kinase inhibitor with IC50s of 132 nM for PDGFRα and 6115 nM for PDGFRβ. It is for research use only and not sold to patients.
  • Highly selective type II PDGFRα kinase inhibitor
  • Exhibits weak inhibition to DDR1 kinase (IC50: 2462±126 nM)
Catalog No. 50-003-44248 Supplier Medchemexpress LLC Supplier No. HY11150750MG
PDGFRα kinase inhibitor 1 is a highly selective type II PDGFRα kinase inhibitor with IC50s of 132 nM for PDGFRα and 6115 nM for PDGFRβ. It is for research use only and not sold to patients.
  • Highly selective type II PDGFRα kinase inhibitor
  • Exhibits weak inhibition to DDR1 kinase (IC50: 2462±126 nM)