Learn More
Carnabio Usa Inc PDGFRA(PDGFRA)(T674I)/5UG/CYT
Supplier:  Carnabio Usa Inc 085035
Encompass
PDGFR(PDGFRA) [T674I], Cytoplasmic domain, Mutant [T674I], Amino Acid 550-1089, NP_006197.1, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0564