Learn More
Sino Biological Recombinant Human PDGF alpha receptor/PDGFRA Protein (ECD, His & AVI Tag), Biotinylated
Supplier:  Sino Biological 10556H27HB20
Encompass_Preferred
A DNA sequence encoding the human PDGFRA (NP_001334758.1) (Met1-Glu524) was expressed with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-211-5227