Learn More
Sino Biological Human PDGFRA / CD140a Protein (His & AVI Tag), Biotinylated 100ug
Supplier:  Sino Biological 10556H27HB100
Encompass_Preferred
A DNA sequence encoding the human PDGFRA (NP_001334758.1) (Met1-Glu524) was expressed with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-211-5260