Learn More
Sino Biological Recombinant Mouse PDGF alpha receptor/PDGFRA Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 57064M08H50
Encompass_Preferred
A DNA sequence encoding the Mouse PDGFRA (NP_001334647.1) (Met1-Ala528) was expressed with a polyhistide tag at the C-terminus.
Catalog No. 50-211-5649