Learn More
ACROBiosystems ACROBiosystems Human PDGF R alpha / PDGFRA Protein, His Tag
Supplier:  ACROBiosystems PDAH52H71MG
Encompass_Preferred
Recombinant Protein;1MG;Human PDGF R alpha, His Tag (PDA-H52H7) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Ala 528 (Accession # P16234-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 58.4 kDa. The protein migrates as 66-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human PDGF-BB, Tag Free (Cat. No. PDB-H4112) at 2 μg/mL (100 μL/well) can bind Human PDGF R alpha, His Tag (Cat. No. PDA-H52H7) with a linear range of 0.02-0.625 μg/mL (QC tested).
Catalog No. 50-195-6859