Learn More
Sino Biological Recombinant Human PDGF beta receptor/PDGFRB Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 10514H02H1100
Encompass_Preferred
A DNA sequence encoding the human PDGFRB (NP_002600.1) (Met1-Phe530) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-211-5068