Learn More
Sino Biological Recombinant Human PDGF beta receptor/PDGFRB Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 10514H08HB100
Encompass_Preferred
A DNA sequence encoding the human PDGFRβ (NP_002600.1) extracellular domain (Met1-Lys531) was expressed, with a C-terminal polyhistidine tag. The expressed protein was biotinylated in vitro.
Catalog No. 50-211-6130