Learn More
Cayman Chemical PDGFRCD140b EXrAClular D 100ug
SDP
Catalog No. 502947005
Encompass_Preferred
Source: Active recombinant human C-terminal His-tagged PDGFRβ expressed in HEK293 cells • Amino acids: 33-531 • MW: 57.6 kDa
Catalog No. 50-294-7005 Supplier Cayman Chemical Supplier No. 33986100 UG
Source: Active recombinant human C-terminal His-tagged PDGFRβ expressed in HEK293 cells • Amino acids: 33-531 • MW: 57.6 kDa