Learn More
Carnabio Usa Inc PDHK3(PDK3)/5UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 101245
Encompass
PDHK3(PDK3), Full-length, Wild type, Amino Acid 1-406, NP_005382.1, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0569