Learn More
PDI™ Castile Soap Towelette
Description
- Contains gentle, pH balanced cleansers
- With 2% coconut oil
Specifications
Specifications
| Product Type | Castile Soap Towelette |
| Form | Wipe |
| Composition | Coconut oil 2% |
| For Use With (Application) | Ideal for midstream clean-catch urine specimen procedures |
| Quantity | 1000/Cs. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.