Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDI™ Hygea™ Sterile Saline Wipes

Catalog No. 2350070
Encompass
Change view
Click to view available options
Quantity:
24 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-500-70 24 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-500-70 Supplier PDI™ Supplier No. C22370
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Help reduce the risk of cross contamination and waste of bottled saline solution

  • Deliver a consistent dose of sterile saline with every application
  • Convenient, cost-effective alternative to bottled saline and gauze pads
  • Non-irritating, mild and gentle cleansing for even the most fragile skin and sensitive skin
  • One step easy-to-use application
  • Saturated with 3mL of sterile saline

Order Info

Wipe size: 6 x 4 in. (15.4 x 10.2cm). Box of 24 individually packaged wipes

Specifications

Product Type Saline Wipes
For Use With (Application) For mild and gentle cleansing, the sterile saline wipes are a great alternative to bottled saline and gauze pads, saturated with 3mL of sterile saline
Quantity 24 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.