Learn More
Enzo Life Sciences PDI monoclonal antibody (1D3) (50µg)
SDP
Supplier:  Enzo Life Sciences ENZABS6860050
Encompass_Preferred
Clone: 1D3. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to the sequence near the C-terminus of rat PDI. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human, Mouse, Rat, Xenopus, Sheep, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Dog, Chicken, Bovine. Applications: ICC, IF, IHC, WB Electron microscopy. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-201-3216