Learn More
Enzo Life Sciences PDI monoclonal antibody (1D3) (PE conjugate) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPA891PEF
Encompass_Preferred
Clone: 1D3. Host: Mouse. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to the sequence near the C-terminus of rat PDI. Source: Purified from ascites. Species reactivity: Human, Mouse, Rat, Xenopus, Sheep, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Dog, Chicken, Bovine. Applications: Flow Cytometry. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide. Handling: Avoid freeze/thaw cycles. Protect from light.
Catalog No. 50-200-7581