Learn More
PDI™ Povidone-Iodine Prep Pad
Catalog No. 0666970
Encompass
Change view
Click to view available options
Size:
Medium
Catalog No. Size
06-669-70 Medium
1 options
This item is not returnable. View return policy
Catalog No. 06-669-70 Supplier PDI™ Supplier No. B40600
Add to Cart
Add to Cart
This item is not returnable. View return policy
Povidone-iodine provides longer germicidal activity than ordinary iodine solution
  • Non-irritating and virtually non-stinging for better patient comfort
  • Perfect for venipuncture, I.V. starts, kidney dialysis, pre-op prep, and other minor invasive procedures
  • Saturated with a 10% USP povidone-iodine prep solution provide proven antisepsis
Order Info
Pad size: 1.1 x 2.6 in. (3 x 6.6cm). 100 individually wrapped pads per box, 10 boxes per case.
Specifications
Product Type PVP Iodine Prep Pad
For Use With (Application) Provides longer germicidal activity than ordinary iodine solution
Size Medium