Learn More
PDI™ Sani-Cloth™ AF Germicidal Disposable Wipes
Catalog No. 23100142
Change view
Click to view available options
Large Canister
Catalog No.
23-100-142 Large Canister
1 options
This item is not returnable. View return policy
Catalog No. 23-100-142 Supplier PDI™ Supplier No. P84372
This item is not returnable. View return policy
Alcohol-free disinfectant wipes are ideal for use on sensitive equipment
  • Designed to make surface disinfection more efficient for staff and pleasant for patients
  • Alcohol- and fragrance-free formula
  • Fragrance free makes it ideal for use around patients and staff, even those with respiratory sensitivities
  • Effective against 44 clinically relevant microoganisms in three minutes, including Multi-Drug Resistant Organisms (MDROs), Bloodborne Pathogens, TB and viruses
  • Compatible with a broad range of hard, non-porous surfaces and equipment commonly found in healthcare environments
Compliance
Meets CDC, OSHA and CMS Tag F441 guidelines.