Learn More
PDI™ Sani-Cloth™ HB Germicidal Disposable Wipes
Catalog No. 0666994
Change view
Click to view available options
Size:
X-Large
Catalog No. Size
06-669-94 X-Large
1 options
This item is not returnable. View return policy
Catalog No. 06-669-94 Supplier PDI™ Supplier No. Q08472
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
Alcohol free, quaternary formula
  • Effective against over 100 organisms in 10 minutes, including: HBV, influenza A/Hong Kong (H3N2), Acinetobacter baumannii, Aspergillus niger, and Trichophyton mentagrophytes
  • Compatible with a broad range of hard, non-porous surfaces and equipment commonly found in healthcare environments
Compliance
Meets CDC, OSHA and CMS Tag F441 guidelines.
Specifications
Certifications/Compliance EPA, CDC, OSHA
Composition 5% Organic soil
Container Type Canister
Disposable Single-use
For Use With (Application) Broad range of hard, non porous surfaces and equipment commonly found in healthcare environments, kills 99.999% of germs on many surfaces throughout fitness centers
Size X-Large
Test Time 10 min.