Learn More
Aviva Systems Biology Corp Pdia3 Antibody - N-terminal region (ARP63564_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63564_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Pdia3. It was validated on Western Blot. . Alternate names: 58kDa, ERp57, ERp60, ERp61, Erp, Grp58, PDI, PDI-Q2, PI-PLC, PLC[a], PlcaApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-288-013