Learn More
Aviva Systems Biology Corp PDIA3 Antibody - C-terminal region (ARP63565_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63565_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PDIA3. It was validated on Western Blot. . Alternate names: ER60, ERp57, ERp60, ERp61, GRP57, GRP58, HsT17083, P58, PI-PLCApplicationIHC, IP, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-288-014