Learn More
Aviva Systems Biology Corp PDIA6 Antibody - middle region (ARP52103_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52103_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PDIA6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PDIA6. Alternate names: ERP5, P5, TXNDC7ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60682