Learn More
Enzo Life Sciences PDIA6 (human) polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ALX210889R100
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to aa 476-492 (D476IDLSDVELDDLGKDEL492) of human PDIA6 (protein disulfide-isomerase A6; protein disulfide isomerase P5). Species reactivity: Human. Applications: Flow Cytometry, WB. Formulation: Liquid. Neat serum containing 0.01% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-8181