Learn More
AdipoGen PDITC
SDP
Supplier:  AdipoGen CDXP0022G100
Encompass_Preferred
Chemical. CAS 4044-65-9. Formula C8H4N2S2. MW 192.26. Synthetic. Reagent for solid-phase sequencing.
Catalog No. 50-256-0272