Learn More
Medchemexpress LLC PDK1 Human sf9 Hi 50ug
SDP
Catalog No. 5000179517
Encompass_Preferred
PDK1 protein is a key kinase that controls glucose and fatty acid metabolism by phosphorylating pyruvate dehydrogenase subunits PDHA1 and PDHA2 This molecular switch inhibits pyruvate dehydrogenase fine-tunes metabolite flux and downregulates aerobic respiration PDK1 Protein Human (sf9 His) is the recombinant human-derived PDK1 protein expressed by Sf9 insect cells with N-His labeled tag
Catalog No. 50-001-79517 Supplier Medchemexpress LLC Supplier No. HYP7371850UG
PDK1 protein is a key kinase that controls glucose and fatty acid metabolism by phosphorylating pyruvate dehydrogenase subunits PDHA1 and PDHA2 This molecular switch inhibits pyruvate dehydrogenase fine-tunes metabolite flux and downregulates aerobic respiration PDK1 Protein Human (sf9 His) is the recombinant human-derived PDK1 protein expressed by Sf9 insect cells with N-His labeled tag