Learn More
Medchemexpress LLC PDK1-IN-RS2 5mg | 1643958-89-7 | 5 MG
SDP
Catalog No. 5000229317
Encompass_Preferred
PDK1-IN-RS2 is a substrate-selective small-molecule inhibitor of 3-phosphoinositide-dependent protein kinase-1 (PDK1). It mimics the peptide docking motif (PIFtide) and is used in biochemical and cellular studies to probe PDK1-dependent signaling within the PI3K/Akt/mTOR pathway.
  • Substrate-selective PDK1 inhibitor (Kd ≈ 9 μM).
  • Useful for studying PI3K/Akt/mTOR pathway and downstream kinase activation.
  • Chemical formula C15H9ClN2O2S3; molecular weight 380.89 g/mol.
  • Available as a 5 mg solid suitable for assay preparation and compound screening.
  • High purity (reported 98.49%) for reliable biochemical results.
Catalog No. 50-002-29317 Supplier Medchemexpress LLC Supplier No. HY1146455MG
PDK1-IN-RS2 is a substrate-selective small-molecule inhibitor of 3-phosphoinositide-dependent protein kinase-1 (PDK1). It mimics the peptide docking motif (PIFtide) and is used in biochemical and cellular studies to probe PDK1-dependent signaling within the PI3K/Akt/mTOR pathway.
  • Substrate-selective PDK1 inhibitor (Kd ≈ 9 μM).
  • Useful for studying PI3K/Akt/mTOR pathway and downstream kinase activation.
  • Chemical formula C15H9ClN2O2S3; molecular weight 380.89 g/mol.
  • Available as a 5 mg solid suitable for assay preparation and compound screening.
  • High purity (reported 98.49%) for reliable biochemical results.