Learn More
Enzo Life Sciences PDK 1 polyclonal antibody (50 µg)
SDP
Supplier:  Enzo Life Sciences ADIKAPPK112D
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of human PDK1 (Pyruvate dehydrogenase kinase-1). UniProt: Q15118. Source: Purified from rabbit serum. Species Reactivity: Human, Mouse, Rat, Rabbit, Porcine, Monkey, Dog. Applications: IHC (PS), WB. Recommended dilutions: Western Blot (1:1,000, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-666-578