Learn More
ECM Bioscience PDK1 Phospho-Regulation Antibody Sampler Kit, ECM Biosciences
Supplier:  ECM Bioscience PK6400
Encompass_Preferred
The PDK1 phospho-regulation antibody sampler kit can be used to examine phosphorylation of PDK1 at Ser-241 and Tyr-9. The kit also includes monoclonal and polyclonal antibodies to monitor total expression levels of PDK1, as well as secondary reagents for antibody detection. Applications: WB, E, ICC; Species Reactivity: Hu, Rt, Ms
Catalog No. 50-206-8778