Learn More
Aviva Systems Biology Corp PDK3 Antibody - N-terminal region (ARP46127_P050) | 100uL
SDP
Catalog No. 5001459238
Encompass_Preferred
This is a rabbit polyclonal antibody against PDK3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PDK3. Alternate names: -ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59238 Supplier Aviva Systems Biology Corp Supplier No. ARP46127_P050
This is a rabbit polyclonal antibody against PDK3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PDK3. Alternate names: -ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish