Learn More
Aviva Systems Biology Corp PDK4 Antibody - N-terminal region (ARP32787_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32787_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PDK4. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human PDK4. Alternate names: FLJ40832ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54764